CELLS HUMAN NTS2 CHO-K1 ARRESTED 11VL
Catalogue # 76847-340
Fournisseur: MULTISPAN INC
CELLS HUMAN NTS2 CHO-K1 ARRESTED 11VL
Catalogue # 76847-340
Fournisseur: MULTISPAN INC
Supplier Number: DC1277-1-11VL
Frequently Bought Together
RNase AWAY® and DNA AWAY™ Surface Decontaminants, Molecular BioProducts
Fournisseur: Thermo Scientific
Acide chlorhydrique 0.10 N analytical reagent, VWR Chemicals BDH®
Fournisseur: VWR International
VWR® Sleeve Stoppers
Fournisseur: VWR International
BD Vacutainer® Venous Blood Collection Tubes
Fournisseur: BD
Orion™ pH Buffer Individual Use Pouches, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Polysorbate 20, Reagent Grade
Fournisseur: VWR
Orion™ pH Buffer Bottles, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Ocean Pacific® Ninja™ Powder-Free Nitrile Medical Examination Gloves, Medicom
Fournisseur: Medicom
Eau, HiPerSolv CHROMANORM for HPLC, VWR Chemicals BDH®
Fournisseur: VWR International
Méthanol ≥99.9%, OmniSolv® LC-MS LC-MS for LC-MS, Supelco®
Fournisseur: MilliporeSigma
Snap Caps for B352 Specimen Bottle, Simport Scientific
Fournisseur: Simport Scientific
Pharmacy Bags, Reclosable, Elkay Plastics
Fournisseur: Elkay Plastics
Bromothymol blue 0.04% (w/v) in aqueous solution indicator
Fournisseur: Ricca Chemical
VWR® Grade 707 Blotting Paper, Grade 707
Fournisseur: VWR International
VWR® Blotting Paper, Grade 703
Fournisseur: VWR International
Sodium perchlorate monohydrate 97+%
Fournisseur: Thermo Scientific Chemicals
Potassium Sulfate
Fournisseur: Ward's Science