CELLS HUMAN GPR43 CHO-K1 ARRESTED 4VL
Catalogue # 76846-092
Fournisseur: MULTISPAN INC
CELLS HUMAN GPR43 CHO-K1 ARRESTED 4VL
Catalogue # 76846-092
Fournisseur: MULTISPAN INC
Supplier Number: DC1104-1-4VL
Frequently Bought Together
COD Digestion vials, VWR Chemicals BDH®
Fournisseur: VWR International
Histoplex™ Histology Containers, Starplex®
Fournisseur: Starplex
pH Reference Standard Buffers, VWR Chemicals BDH®
Fournisseur: BDH
Masterflex® I/P® Spooled Pump Tubing, Platinum-Cured Silicone, Avantor®
Fournisseur: VWR International
BD Vacutainer® Venous Blood Collection Tubes
Fournisseur: BD
Acétonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
Leakbuster™ Specimen Containers
Fournisseur: Starplex
Formalin 10%, neutral buffered, VWR®
Fournisseur: VWR International
VACUETTE® Secondary Tubes, Greiner Bio-One
Fournisseur: Greiner Bio-One
Components and Accessories for Miele Lab Washers, Miele Professional
Fournisseur: Miele
VWR® Premium Dissecting Scissors with Sharp/Blunt Blades
Fournisseur: VWR International
HardyVal™ Tryptic Soy Broth, USP, Media-Bag™, Hardy Diagnostics
Fournisseur: Hardy Diagnostics
Enzymes and Separation Accessories, Omega Bio-Tek
Fournisseur: Omega Bio-Tek
Sodium Phosphate Dibasic Anhydrous
Fournisseur: Ward's Science
N,N-Diméthylacétamide ≥99.9%, OmniSolv®, Supelco®
Fournisseur: MilliporeSigma
4-Methylpiperidine 98+%
Fournisseur: Thermo Scientific Chemicals