Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
VWR® One-Step RT-qPCR Kit | Avantor

VWR® One-Step RT-qPCR Kit

VWR® One-Step RT-qPCR kit, 1000 (2x 500)
Catalog # 73212-1000
Supplier:   VWR International
VWR® One-Step RT-qPCR Kit
Catalog # 73212-1000
Supplier:  VWR International
CAS Number:  
VWR® One-Step RT-qPCR Kit
VWR® One-Step RT-qPCR Kit

VWR® One-Step RT-qPCR kit, 1000 (2x 500)

CAS Number:  
Price
$1,568.86
Log in pour la disponibilité
Each (1,000Assays)
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

VWR® One-Step RT-qPCR Kit is designed to facilitate rapid, sensitive, and precise detection and quantification of various RNA sequences via dye-based qPCR. It features a distinctive universal passive reference dye that is compatible with ROX-dependent and ROX-independent qPCR instruments.

  • For the detection and quantification of RNA targets using the SYBR®/FAM channel
  • Ready-to-use kit containing all necessary reaction components, except for primers and RNA template
  • Suitable for a variety of RNA sample types
  • Includes Hot Start Taq DNA Polymerase and VWR® Reverse Transcriptase for improved reaction specificity
  • Incorporates dUTP for carryover contamination prevention
  • VWR® One-Step Reaction Mix is supplied at a 2X concentration

The VWR® RT Enzyme Mix, supplied at a 20X concentration, contains the specially designed, temperature-controlled VWR® Reverse Transcriptase and Murine RNase Inhibitor to assist in preventing RNA degradation. This temperature-controlled RT enzyme prevents unwanted non-specific activity and displays superior thermostability in comparison to other reverse transcriptases.

Caution: Store at -20°C. For research use only.

Spécifications

Spécifications

  • Désignation:
    VWR® One-Step RT-qPCR kit
  • No. of Reactions:
    1000 (2x 500)
  • Cat. No.:
    73212-1000
  • One step:
    Yes
  • Application:
    RT-qPCR dye

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.