Optimum Growth® Flasks
Optimum Growth™ flask, double bagged, 1.6 ml, Translucent, Sterile, Screw cap
Catalog # 76808-418
 :  931113-DB
Optimum Growth® Flasks
Catalog # 76808-418
Optimum Growth® Flasks
Optimum Growth® Flasks
Optimum Growth™ flask, double bagged, 1.6 ml, Translucent, Sterile, Screw cap
Price
$794.00
Log in pour la disponibilité
Case of 12
Product Details & Documents
Thomson’s patented Optimum Growth® Flasks are engineered for mammalian and insect cell culture, supporting applications from research to production. Available in a wide variety of sizes, these flasks support 40 to 50% fill capacity (vs. 33% in traditional shake flasks) and maintain consistent performance across scales, making them a leading choice for modern bioprocessing workflows.
  • Higher culture volume per flask helps grow more cells in the same shaker space, increasing throughput
  • Same footprint as standard Fernbach flasks, but with greater media capacity and less foaming
  • Unique low shear baffle design increases aeration while preserving cell viability
  • 0.2 μm PTFE vented caps maintain sterility and efficient gas exchange
  • Transfer cap options connect directly to cell bags or bioreactors (quick-connect, luer lock, or tube fusion)
  • Scalable across all sizes; supporting consistent shake speeds and growth conditions from 125 ml to 7 L
  • Easier to handle than bioreactor bags or bioprocess containers; no rocker bioreactor, pump, or complex tubing setup required
  • Individually packaged, double-bagged, and sterilized for cleanroom-ready use
  • Made in the USA with most products available for same-day shipping
The Optimum Growth® System also includes transfer caps and the Rapid Clear® Cap for clarification, offering a modular and cGMP-compliant platform for high-yield, scalable bioprocessing.
Spécifications
  • Cap Type:
    Screw cap
  • Sterility:
    Sterile
  • Capacité:
    1.6 ml
  • Color:
    Translucent
  • Désignation:
    Optimum Growth™ flask, double bagged
Accessories
Informations sur le produit
  • Catalog #
    Material
    Connection
    Availability
    Price
  • Inversion transfer cap for 1.6 and 2.8 L flasks, with 0.2 µm PTFE vent, C-Flex® 24 tubing (⁷/₁₆"Ext.Ø.׳/₁₆"Int.Ø)
    Inversion transfer cap for 1.6 and 2.8 L flasks, with 0.2 µm PTFE vent, C-Flex® 24 tubing (⁷/₁₆"Ext.Ø.׳/₁₆"Int.Ø)
    76808-342
    Polypropylene
    Tube fuse (plug on terminus)
    Log in pour la disponibilité
    Case of 4 - $885.00
Informations sur le produit
  • Catalog #
    Ø
    Availability
    Price
  • Stand with ring for inverting 1.6 and 2.8 L Optimum Growth® flasks
    Stand with ring for inverting 1.6 and 2.8 L Optimum Growth® flasks
    76808-514
    5" ring
    Log in pour la disponibilité
    Each - $340.00
Informations sur le produit
  • Catalog #
    Sterility
    Availability
    Price
  • Optimum Growth® vent caps for 250 ml flasks
    Optimum Growth® vent caps for 250 ml flasks
    76808-546
    Sterile
    Log in pour la disponibilité
    Case of 50 - $641.00
Informations sur le produit
  • Catalog #
    Material
    Connection
    Availability
    Price
  • Inversion transfer cap for 5 L flasks, with 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)
    Inversion transfer cap for 5 L flasks, with 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)
    76808-348
    Polypropylene
    Male Luer Lock, tube fuse
    Log in pour la disponibilité
    Case of 4 - $1,030.00
Informations sur le produit
  • Catalog #
    Material
    Fits
    Availability
    Price
  • Bi-directional transfer cap for 1.6 L flasks, with threaded top, 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)
    Bi-directional transfer cap for 1.6 L flasks, with threaded top, 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)
    76808-322
    Polypropylene
    1.6 L flask
    Log in pour la disponibilité
    Case of 8 - $1,770.00
Informations sur le produit
  • Catalog #
    Ø
    Availability
    Price
  • Ring only for inverting 5 L Optimum Growth® flasks
    Ring only for inverting 5 L Optimum Growth® flasks
    76808-512
    7" ring
    Log in pour la disponibilité
    Each - $170.00
Informations sur le produit
  • Catalog #
    Material
    Fits
    Availability
    Price
  • Bi-directional transfer cap for 2.8 L flasks, with threaded top, 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)
    Bi-directional transfer cap for 2.8 L flasks, with threaded top, 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)
    76808-326
    Polypropylene
    2.8 L
    Log in pour la disponibilité
    Case of 8 - $1,770.00
Informations sur le produit
  • Catalog #
    Material
    Connection
    Availability
    Price
  • Inversion transfer cap for 5 L flasks, with 0.2 µm PTFE vent
    Inversion transfer cap for 5 L flasks, with 0.2 µm PTFE vent
    76808-332
    Polypropylene
    ¹/₄" Ext.Ø. barbed quick connect
    Log in pour la disponibilité
    Case of 4 - $734.00
Informations sur le produit
  • Catalog #
    Ø
    Availability
    Price
  • Ring only for inverting 1.6 and 2.8 L Optimum Growth® flasks
    Ring only for inverting 1.6 and 2.8 L Optimum Growth® flasks
    76808-516
    5" ring
    Log in pour la disponibilité
    Each - $170.00
Informations sur le produit
  • Catalog #
    Material
    Connection
    Availability
    Price
  • Inversion transfer cap for 1.6 and 2.8 L flasks, with 0.2 µm PTFE vent
    Inversion transfer cap for 1.6 and 2.8 L flasks, with 0.2 µm PTFE vent
    76808-334
    Polypropylene
    ⁷/₁₆" Ext.Ø. male quick connect
    Log in pour la disponibilité
    Case of 4 - $605.00
Informations sur le produit
  • Catalog #
    Material
    Connection
    Availability
    Price
  • Inversion transfer cap for 1.6 and 2.8 L flasks, with 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)
    Inversion transfer cap for 1.6 and 2.8 L flasks, with 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)
    76808-346
    Polypropylene
    Male Luer Lock, tube fuse
    Log in pour la disponibilité
    Case of 4 - $885.00
Informations sur le produit
  • Catalog #
    Dimensions
    Availability
    Price
  • 8-position carrier for 250 ml Optimum Growth® flasks
    8-position carrier for 250 ml Optimum Growth® flasks
    76808-362
    13.4×6"
    Log in pour la disponibilité
    Each - $1,210.00
Informations sur le produit
  • Catalog #
    Sterility
    Availability
    Price
  • Optimum Growth® vent caps for 125 ml flasks
    Optimum Growth® vent caps for 125 ml flasks
    76808-544
    Sterile
    Log in pour la disponibilité
    Case of 50 - $641.00
Informations sur le produit
  • Catalog #
    Sterility
    Availability
    Price
  • Optimum Growth® vent caps for 1.6 L, 2.8 L, and 5 L flasks
    Optimum Growth® vent caps for 1.6 L, 2.8 L, and 5 L flasks
    76808-554
    Sterile
    Log in pour la disponibilité
    Case of 24 - $603.00
Informations sur le produit
  • Catalog #
    Sterility
    Availability
    Price
  • Optimum Growth® vent caps for 500 ml flasks
    Optimum Growth® vent caps for 500 ml flasks
    76808-548
    Sterile
    Log in pour la disponibilité
    Case of 25 - $387.00
Informations sur le produit
  • Catalog #
    Material
    Connection
    Availability
    Price
  • Inversion transfer cap for 1.6 and 2.8 L flasks, with 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)
    Inversion transfer cap for 1.6 and 2.8 L flasks, with 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)
    76808-338
    Polypropylene
    Tube fuse (plug on terminus)
    Log in pour la disponibilité
    Case of 4 - $853.00
Informations sur le produit
  • Catalog #
    Material
    Fits
    Availability
    Price
  • Bi-directional transfer cap for 1.6 L flasks, with threaded top, 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)
    Bi-directional transfer cap for 1.6 L flasks, with threaded top, 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)
    76808-324
    Polypropylene
    1.6 L flask
    Log in pour la disponibilité
    Case of 8 - $1,770.00
Informations sur le produit
  • Catalog #
    Material
    Sterility
    Availability
    Price
  • Rapid Clear® cap 3000
    Rapid Clear® cap 3000
    76808-350
    Polypropylene
    Sterile
    Log in pour la disponibilité
    Case of 4 - $1,700.00
Informations sur le produit
  • Catalog #
    Dimensions
    Availability
    Price
  • 8-position carrier for 125 ml Optimum Growth® flasks
    8-position carrier for 125 ml Optimum Growth® flasks
    76808-360
    10.75×5"
    Log in pour la disponibilité
    Each - $1,210.00
Informations sur le produit
  • Catalog #
    Sterility
    Availability
    Price
  • Optimum Growth® 1.6 L flask and bioreactor exhaust caps
    Optimum Growth® 1.6 L flask and bioreactor exhaust caps
    76808-330
    Sterile
    Log in pour la disponibilité
    Case of 8 - $2,210.00
Informations sur le produit
  • Catalog #
    Material
    Connection
    Availability
    Price
  • Inversion transfer cap for 5 L flasks, with 0.2 µm PTFE vent
    Inversion transfer cap for 5 L flasks, with 0.2 µm PTFE vent
    76808-336
    Polypropylene
    ⁷/₁₆" Ext.Ø. male quick connect
    Log in pour la disponibilité
    Case of 4 - $605.00
Informations sur le produit
  • Catalog #
    Sterility
    Availability
    Price
  • Optimum Growth® low evaporation caps for 1.6, 2.8, and 5 L flasks
    Optimum Growth® low evaporation caps for 1.6, 2.8, and 5 L flasks
    76808-354
    Sterile
    Log in pour la disponibilité
    Case of 24 - $415.00
Informations sur le produit
  • Catalog #
    Material
    Fits
    Availability
    Price
  • Bi-directional transfer cap for 5 L flasks, with threaded top, 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)
    Bi-directional transfer cap for 5 L flasks, with threaded top, 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)
    76808-328
    Polypropylene
    5 L flask
    Log in pour la disponibilité
    Case of 8 - $1,770.00
Informations sur le produit
  • Catalog #
    Material
    Connection
    Availability
    Price
  • Inversion transfer cap for 5 L flasks, with 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)
    Inversion transfer cap for 5 L flasks, with 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)
    76808-340
    Polypropylene
    Tube fuse (plug on terminus)
    Log in pour la disponibilité
    Case of 4 - $885.00
Informations sur le produit
  • Catalog #
    Material
    Fits
    Availability
    Price
  • Bi-directional transfer cap for 5 L flasks, with threaded top, 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)
    Bi-directional transfer cap for 5 L flasks, with threaded top, 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)
    76808-320
    Polypropylene
    5 L flask
    Log in pour la disponibilité
    Case of 8 - $1,770.00
Informations sur le produit
  • Catalog #
    Material
    Connection
    Availability
    Price
  • Inversion transfer cap for 5 L flasks, with 0.2 µm PTFE vent, C-Flex® 24 tubing (⁷/₁₆"Ext.Ø.׳/₁₆"Int.Ø)
    Inversion transfer cap for 5 L flasks, with 0.2 µm PTFE vent, C-Flex® 24 tubing (⁷/₁₆"Ext.Ø.׳/₁₆"Int.Ø)
    76808-344
    Polypropylene
    Tube fuse (plug on terminus)
    Log in pour la disponibilité
    Case of 4 - $885.00
Informations sur le produit
  • Catalog #
    Ø
    Availability
    Price
  • Stand with ring for inverting 5 L Optimum Growth® flasks
    Stand with ring for inverting 5 L Optimum Growth® flasks
    76808-510
    7" ring
    Log in pour la disponibilité
    Each - $340.00