Optimum Growth® Flasks
Product Details & Documents
Thomson’s patented Optimum Growth® Flasks are engineered for mammalian and insect cell culture, supporting applications from research to production. Available in a wide variety of sizes, these flasks support 40 to 50% fill capacity (vs. 33% in traditional shake flasks) and maintain consistent performance across scales, making them a leading choice for modern bioprocessing workflows.
Higher culture volume per flask helps grow more cells in the same shaker space, increasing throughput
- Same footprint as standard Fernbach flasks, but with greater media capacity and less foaming
- Unique low shear baffle design increases aeration while preserving cell viability
- 0.2 μm PTFE vented caps maintain sterility and efficient gas exchange
- Transfer cap options connect directly to cell bags or bioreactors (quick-connect, luer lock, or tube fusion)
- Scalable across all sizes; supporting consistent shake speeds and growth conditions from 125 ml to 7 L
- Easier to handle than bioreactor bags or bioprocess containers; no rocker bioreactor, pump, or complex tubing setup required
- Individually packaged, double-bagged, and sterilized for cleanroom-ready use
- Made in the USA with most products available for same-day shipping
The Optimum Growth® System also includes transfer caps and the Rapid Clear® Cap for clarification, offering a modular and cGMP-compliant platform for high-yield, scalable bioprocessing.
Spécifications
- Cap Type:Screw cap
- Sterility:Sterile
- Capacité:5 L
- Color:Translucent
- Désignation:Optimum Growth™ flask, double bagged
Accessories
Informations sur le produit
- Catalog #MaterialConnectionAvailabilityPrice
- Inversion transfer cap for 1.6 and 2.8 L flasks, with 0.2 µm PTFE vent, C-Flex® 24 tubing (⁷/₁₆"Ext.Ø.׳/₁₆"Int.Ø)Inversion transfer cap for 1.6 and 2.8 L flasks, with 0.2 µm PTFE vent, C-Flex® 24 tubing (⁷/₁₆"Ext.Ø.׳/₁₆"Int.Ø)76808-342PolypropyleneTube fuse (plug on terminus)Log in pour la disponibilitéCase of 4 - $885.00
Informations sur le produit
- Catalog #ØAvailabilityPrice
- Stand with ring for inverting 1.6 and 2.8 L Optimum Growth® flasksStand with ring for inverting 1.6 and 2.8 L Optimum Growth® flasks76808-5145" ringLog in pour la disponibilitéEach - $340.00
Informations sur le produit
- Catalog #SterilityAvailabilityPrice
- Optimum Growth® vent caps for 250 ml flasksOptimum Growth® vent caps for 250 ml flasks76808-546SterileLog in pour la disponibilitéCase of 50 - $641.00
Informations sur le produit
- Catalog #MaterialConnectionAvailabilityPrice
- Inversion transfer cap for 5 L flasks, with 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)Inversion transfer cap for 5 L flasks, with 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)76808-348PolypropyleneMale Luer Lock, tube fuseLog in pour la disponibilitéCase of 4 - $1,030.00
Informations sur le produit
- Catalog #MaterialFitsAvailabilityPrice
- Bi-directional transfer cap for 1.6 L flasks, with threaded top, 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)Bi-directional transfer cap for 1.6 L flasks, with threaded top, 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)76808-322Polypropylene1.6 L flaskLog in pour la disponibilitéCase of 8 - $1,770.00
Informations sur le produit
- Catalog #ØAvailabilityPrice
- Ring only for inverting 5 L Optimum Growth® flasksRing only for inverting 5 L Optimum Growth® flasks76808-5127" ringLog in pour la disponibilitéEach - $170.00
Informations sur le produit
- Catalog #MaterialFitsAvailabilityPrice
- Bi-directional transfer cap for 2.8 L flasks, with threaded top, 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)Bi-directional transfer cap for 2.8 L flasks, with threaded top, 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)76808-326Polypropylene2.8 LLog in pour la disponibilitéCase of 8 - $1,770.00
Informations sur le produit
- Catalog #MaterialConnectionAvailabilityPrice
- Inversion transfer cap for 5 L flasks, with 0.2 µm PTFE ventInversion transfer cap for 5 L flasks, with 0.2 µm PTFE vent76808-332Polypropylene¹/₄" Ext.Ø. barbed quick connectLog in pour la disponibilitéCase of 4 - $734.00
Informations sur le produit
- Catalog #ØAvailabilityPrice
- Ring only for inverting 1.6 and 2.8 L Optimum Growth® flasksRing only for inverting 1.6 and 2.8 L Optimum Growth® flasks76808-5165" ringLog in pour la disponibilitéEach - $170.00
Informations sur le produit
- Catalog #MaterialConnectionAvailabilityPrice
- Inversion transfer cap for 1.6 and 2.8 L flasks, with 0.2 µm PTFE ventInversion transfer cap for 1.6 and 2.8 L flasks, with 0.2 µm PTFE vent76808-334Polypropylene⁷/₁₆" Ext.Ø. male quick connectLog in pour la disponibilitéCase of 4 - $605.00
Informations sur le produit
- Catalog #MaterialConnectionAvailabilityPrice
- Inversion transfer cap for 1.6 and 2.8 L flasks, with 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)Inversion transfer cap for 1.6 and 2.8 L flasks, with 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)76808-346PolypropyleneMale Luer Lock, tube fuseLog in pour la disponibilitéCase of 4 - $885.00
Informations sur le produit
- Catalog #DimensionsAvailabilityPrice
- 8-position carrier for 250 ml Optimum Growth® flasks8-position carrier for 250 ml Optimum Growth® flasks76808-36213.4×6"Log in pour la disponibilitéEach - $1,210.00
Informations sur le produit
- Catalog #SterilityAvailabilityPrice
- Optimum Growth® vent caps for 125 ml flasksOptimum Growth® vent caps for 125 ml flasks76808-544SterileLog in pour la disponibilitéCase of 50 - $641.00
Informations sur le produit
- Catalog #SterilityAvailabilityPrice
- Optimum Growth® vent caps for 1.6 L, 2.8 L, and 5 L flasksOptimum Growth® vent caps for 1.6 L, 2.8 L, and 5 L flasks76808-554SterileLog in pour la disponibilitéCase of 24 - $603.00
Informations sur le produit
- Catalog #SterilityAvailabilityPrice
- Optimum Growth® vent caps for 500 ml flasksOptimum Growth® vent caps for 500 ml flasks76808-548SterileLog in pour la disponibilitéCase of 25 - $387.00
Informations sur le produit
- Catalog #MaterialConnectionAvailabilityPrice
- Inversion transfer cap for 1.6 and 2.8 L flasks, with 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)Inversion transfer cap for 1.6 and 2.8 L flasks, with 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)76808-338PolypropyleneTube fuse (plug on terminus)Log in pour la disponibilitéCase of 4 - $853.00
Informations sur le produit
- Catalog #MaterialFitsAvailabilityPrice
- Bi-directional transfer cap for 1.6 L flasks, with threaded top, 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)Bi-directional transfer cap for 1.6 L flasks, with threaded top, 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)76808-324Polypropylene1.6 L flaskLog in pour la disponibilitéCase of 8 - $1,770.00
Informations sur le produit
- Catalog #MaterialSterilityAvailabilityPrice
- Rapid Clear® cap 3000Rapid Clear® cap 300076808-350PolypropyleneSterileLog in pour la disponibilitéCase of 4 - $1,700.00
Informations sur le produit
- Catalog #DimensionsAvailabilityPrice
- 8-position carrier for 125 ml Optimum Growth® flasks8-position carrier for 125 ml Optimum Growth® flasks76808-36010.75×5"Log in pour la disponibilitéEach - $1,210.00
Informations sur le produit
- Catalog #SterilityAvailabilityPrice
- Optimum Growth® 1.6 L flask and bioreactor exhaust capsOptimum Growth® 1.6 L flask and bioreactor exhaust caps76808-330SterileLog in pour la disponibilitéCase of 8 - $2,210.00
Informations sur le produit
- Catalog #MaterialConnectionAvailabilityPrice
- Inversion transfer cap for 5 L flasks, with 0.2 µm PTFE ventInversion transfer cap for 5 L flasks, with 0.2 µm PTFE vent76808-336Polypropylene⁷/₁₆" Ext.Ø. male quick connectLog in pour la disponibilitéCase of 4 - $605.00
Informations sur le produit
- Catalog #SterilityAvailabilityPrice
- Optimum Growth® low evaporation caps for 1.6, 2.8, and 5 L flasksOptimum Growth® low evaporation caps for 1.6, 2.8, and 5 L flasks76808-354SterileLog in pour la disponibilitéCase of 24 - $415.00
Informations sur le produit
- Catalog #MaterialFitsAvailabilityPrice
- Bi-directional transfer cap for 5 L flasks, with threaded top, 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)Bi-directional transfer cap for 5 L flasks, with threaded top, 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)76808-328Polypropylene5 L flaskLog in pour la disponibilitéCase of 8 - $1,770.00
Informations sur le produit
- Catalog #MaterialConnectionAvailabilityPrice
- Inversion transfer cap for 5 L flasks, with 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)Inversion transfer cap for 5 L flasks, with 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)76808-340PolypropyleneTube fuse (plug on terminus)Log in pour la disponibilitéCase of 4 - $885.00
Informations sur le produit
- Catalog #MaterialFitsAvailabilityPrice
- Bi-directional transfer cap for 5 L flasks, with threaded top, 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)Bi-directional transfer cap for 5 L flasks, with threaded top, 0.2 µm PTFE vent, C-Flex® 16 tubing (¹/₈"Int.Ø×¹/₄"Ext.Ø.)76808-320Polypropylene5 L flaskLog in pour la disponibilitéCase of 8 - $1,770.00
Informations sur le produit
- Catalog #MaterialConnectionAvailabilityPrice
- Inversion transfer cap for 5 L flasks, with 0.2 µm PTFE vent, C-Flex® 24 tubing (⁷/₁₆"Ext.Ø.׳/₁₆"Int.Ø)Inversion transfer cap for 5 L flasks, with 0.2 µm PTFE vent, C-Flex® 24 tubing (⁷/₁₆"Ext.Ø.׳/₁₆"Int.Ø)76808-344PolypropyleneTube fuse (plug on terminus)Log in pour la disponibilitéCase of 4 - $885.00
Informations sur le produit
- Catalog #ØAvailabilityPrice
- Stand with ring for inverting 5 L Optimum Growth® flasksStand with ring for inverting 5 L Optimum Growth® flasks76808-5107" ringLog in pour la disponibilitéEach - $340.00