FILTER SAPPHIRE FL 452/45 NM
Catalogue # 76716-990
Fournisseur: Azure Biosystems
FILTER SAPPHIRE FL 452/45 NM
Catalogue # 76716-990
Fournisseur: Azure Biosystems
Supplier Number: IS4008
Frequently Bought Together
Water, distilled
Fournisseur: GROWCELLS
Difco™ Granulated Agar, BD Biosciences
Fournisseur: BD
Sodium hydroxide 5 N in aqueous solution, Biotech Reagent, J.T.Baker®
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
VWR® Absorbent Pads
Fournisseur: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Fournisseur: BDH
B352 Specimen Bottles and Caps, Simport Scientific
Fournisseur: Simport Scientific
Macrosep® Advance Centrifugal Devices, Cytiva (Formerly Pall Lab)
Fournisseur: Cytiva
N,N-Dimethylformamide ≥99.8% (by GC), GR ACS, Supelco®
Fournisseur: MilliporeSigma
Phalloidin Conjugates, Biotium
Fournisseur: Biotium
Hematoxylin modified solution Gill III used in nuclear staining, Sigma-Aldrich®
Fournisseur: MilliporeSigma
STORAGE SOLUTION PH ELECTRODE
Fournisseur: Oakton
Sodium chloride 0.9% in aqueous solution normal saline solution, sterile
Fournisseur: Teknova
Salicylaldehyde azine 97%
Fournisseur: Thermo Scientific Chemicals
Lugol's iodine solution, HARLECO® for Gram staining, Sigma-Aldrich®
Fournisseur: MilliporeSigma
Diversey™ Oxivir® Tb Disinfectant Solution, Antylia Scientific
Fournisseur: Antyila Scientific
TOTAL CHLORINE IND SOL 473ML
Fournisseur: Hach