WHEATON® Serum Vials
Serum vial, 2 ml, amber EP, tray, 2 ml, 14.75×32 mm, Glass, Amber, Non
Supplier #: WUBB2MLA-T-EP
Serum vials converted from Type I borosilicate glass tubing.
- Shrink-wrapped modules reduce particulate contamination
- Special design adds strength for freeze-drying applications
- Crimp cap type
Vials conform to the hydrolytic resistance requirements outlined in USP <660> and EP 3.2.1.
- Avec bouchon:Non
- Capacité:2 ml
- Color:Amber
- Désignation:Serum vial, 2 ml, amber EP, tray
- Matière:Glass
- Diameter ×H:14.75×32 mm



