VWR® Sample Bottles
Product Details & Documents
VWR® sample bottles are ideal for storage of media, laboratory reagents, and biological buffers in temperatures ranging from −70 to 60 °C. Eliminate washing, depyrogenation, and autoclaving steps with sterile and non-pyrogenic bottles.
- Made from durable, break-resistant 100% virgin PETG material
- Suitable for frozen storage down to −70°C
- Heat-shrink band around closure and neck for tamper-evident seal
- Leakproof bottles and HDPE closures provided
- Leakproof sealing with integrated lip seal feature
- Sterile and non-pyrogenic, SAL 10⁻⁶
- Low extractables
- Transparency and reduced permeability
Bottles are arranged in shrink-wrapped trays and polybag packaging to prevent contamination. Bottles 30 ml and above feature volume graduations. Non sterile bottles are autoclavable.
Certifications: US FDA 21CRF, USP Class VI compliant resin.
Informations sur les accessoires: Spare closures are available for specific application requirements.
Packaging: Bottles are packed with band, tray arranged, and poly bagged. Spare closures are bulk packed.
Spécifications
- Taille du bouchon:20-415
- Sterility:Sterile
- Capacité:5 ml
- Matière:PETG
- Taille:Ø 22.5 mm
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



