Student Dissection Kits | Avantor
Student Dissection Kits
Student dissection kit with soft case
Catalog # 470350-634
Supplier:   AFTAB AHMED SE
Fournisseur #:  10-PMVK-1
Student Dissection Kits
Catalog # 470350-634
Supplier:  AFTAB AHMED SE
Student Dissection Kits
Student Dissection Kits
Student dissection kit with soft case
Price
$23.90
Log in pour la disponibilité
Kit of 1 Case of 50
Product Details & Documents
Pre-built instrument kits.
  • Three storage options
  • Easily store between uses
  • Quick setup for student groups
  • Easy instrument check when finished
These three dissection sets provide varied options for what instruments and storage best fit the class. Soft cases are lighter and a little easier to store.
The hard clamshell case provides the most protection from possible sliding instruments, while closing securely. Zippered cases provide storage for a greater variety of instruments, and when closed prevent the instruments from falling out if they weren't securely replaced.
Each set contains different instruments, see attached document for specific contents, with all containing scalpels, scissors, forceps, and teasing needles.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Spécifications
  • Désignation:
    Student dissection kit with soft case
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.