Cardboard Spectrometer Kit
Cardboard spectrometer kit, HS
Catalog # 470350-102
Supplier: Science First
Fournisseur #: 654-0025
Jump to:
Product Details & Documents
Investigate light spectra with this easy-to-use spectrometer.
Recommended for astronomy, chemistry, physics, physical science and earth science
Easy to use
Offers a clear view of the light’s properties
This spectrometer is designed for educational purposes, helping students explore and understand light spectra. Observe absorption lines in the Sun's spectrum, the spectra of energized elements in gaseous states and the double yellow lines of Mercury vapors in the spectrum of a fluorescent lamp.
Spécifications
- Désignation:Cardboard spectrometer kit
- Grade:HS
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.