Elevated Zero Mazes
Elevated Zero Mazes
Elevated zero maze; rat, 100 cm, 10 cm, 30 cm (wall), 61 cm (stand)
Catalog # 76670-000
Supplier:   MAZE ENGINEERS
Supplier #:  4002
Elevated Zero Mazes
Catalog # 76670-000
Supplier:  MAZE ENGINEERS
Elevated Zero Mazes
Elevated Zero Mazes
Elevated zero maze; rat, 100 cm, 10 cm, 30 cm (wall), 61 cm (stand)
Price
$4,940.06
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The elevated zero maze is an elevated, ring-shaped runway with the same amount of area devoted to adjacent open and closed quadrants, similar to the elevated plus maze to study anxiety behaviors.
  • Acrylic construction
  • Odor-free
  • Matted finish for non-reflective surfaces
  • Easy to clean
  • Easy assembly and disssembly with a removable base
The elevated zero maze is very similar to the elevated plus maze, but with the center region of the elevated plus maze removed. It has been pharmacologically validated with various anxiolytic drugs.
The design is created with an elevated circular platform with two walled, enclosed portions as well as two open portions, allowing for clear interpretations of murine behavior. Because mice tend to avoid open-spaced areas, the animals will preferentially spend more time in the enclosed walled area.
Spécifications
  • Ø:
    100 cm
  • Largeur:
    10 cm
  • Désignation:
    Elevated zero maze; rat
  • Hauteur:
    30 cm (wall), 61 cm (stand)