Dispenser Dropping Bottles | Avantor
Dispenser Dropping Bottles
Dispenser dropping bottle, 250 ml, LDPE
Catalog # 470349-768
Fournisseur #:  UNDDB250
Dispenser Dropping Bottles
Catalog # 470349-768
Dispenser Dropping Bottles
Dispenser Dropping Bottles
Dispenser dropping bottle, 250 ml, LDPE
Price
$32.85
Log in pour la disponibilité
Pack of 12
Product Details & Documents
Precision dispensing of solutions.
  • Dispenses one drop at a time
  • Spout lid is connected to the cap
  • LDPE with leakproof PP cap
  • BPA-free
This LDPE dispensing bottle can be easily operated with one hand. The bottle features a polypropylene screw cap with a tethered spout lid that snaps securely in place.
Spécifications
  • Capacité:
    250 ml
  • Désignation:
    Dispenser dropping bottle
  • Matière:
    LDPE