Dropping Bottle
125 ml, LDPE
Catalog # 470349-782
Supplier: United Scientific Supplies
Supplier #: PDB125PK12
Jump to:
Product Details & Documents
Dispense solutions accurately, one drop at a time.
- Ideal for storing and dispensing small volumes
- Translucent design
- LDPE with leakproof PP cap
- BPA-free
Dropping bottles are useful for adding small amounts of a liquid. With a small squeeze the amount of liquid being dispensed can be controlled. Manufactured from translucent and low density polyethylene. Caps are made from polypropylene. Assembled bottles are sold as packs.
Specifications
- Capacité:125 ml
- Matière:LDPE