Class II Laser Pointers
Class II Laser, green
  470349-618
 :  GLPCLS2
Class II Laser Pointers
  470349-618
Class II Laser Pointers
Class II Laser Pointers
Class II Laser, green
$64.15
Each
Class 2 laser are safe for classroom use.
  • Maximum output of less than 1 mW
  • Push-button switch
These economical laser pointers are useful for many optical experiments. Their lower power output is ideal for middle school and high school classrooms. The lasers operate on a push-button switch, are housed in a rubberized aluminum pen-style case, and include a pocket clip.
The 6" long × 0.5" diameter Class 2 Red laser pointer has a maximum output of 1 mW and a wavelength of 650 nm (±10 nm). Range of beam is 75 meters. Operates on two 'AAA' batteries (included).
The 6" long × 0.5" diameter Class 2 Green laser pointer has a maximum output of 1 mW and a wavelength of 532 nm (±10 nm). Range of beam is 125 meters. Operates on two 'AAA' batteries (included).
  • Désignation:
    Class II Laser, green