Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Medium Grade, 15 μm

Medium Grade, 15 μm

Filter Paper, Medium Grade, 320 mm diameter
Catalog # 470349-756
Supplier #:  FPR320
Medium Grade, 15 μm
Catalog # 470349-756
Medium Grade, 15 μm
Medium Grade, 15 μm

Filter Paper, Medium Grade, 320 mm diameter

Price
$52.95
Log in for availability
Pack of 100
Product Details & Documents

The most widely used qualitative filter paper for routine applications.

  • Medium grade, 15 μm

Holds flocculants and precipitates commonly encountered in laboratory exercises.

Packaging: Packages of 100.

Specifications
  • Désignation:
    Filter Paper, Medium Grade, 320 mm diameter