Dispenser Dropping Bottles
Dispenser dropping bottle, 15 ml, LDPE
Catalog # 470349-746
Supplier: United Scientific Supplies
: UNDDB015
Jump to:
Product Details & Documents
Precision dispensing of solutions.
- Dispenses one drop at a time
- Spout lid is connected to the cap
- LDPE with leakproof PP cap
- BPA-free
This LDPE dispensing bottle can be easily operated with one hand. The bottle features a polypropylene screw cap with a tethered spout lid that snaps securely in place.
Spécifications
- Capacité:15 ml
- Désignation:Dispenser dropping bottle
- Matière:LDPE