TE Buffers 0.25X
250 ml
Catalog # CA76645-340
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Supplier #: DNF-497-0250
Jump to:
Product Details & Documents
TE buffers for rinsing capillary arrays.
Spécifications
Spécifications
- Volume:250 ml