TE Buffers 0.25X
TE Buffers 0.25X
125 ml
Catalog # CA76645-140
Supplier #:  DNF-497-0125
TE Buffers 0.25X
Catalog # CA76645-140
CAS Number:  
TE Buffers 0.25X
TE Buffers 0.25X
125 ml
CAS Number:  
Price
$232.00
Log in pour la disponibilité
Each
Minimum order quantity is 1
Product Details & Documents
TE buffers for rinsing capillary arrays.
Spécifications
Spécifications
  • Volume:
    125 ml