Refracting Telescope Kit
Product Details & Documents
This simple lab enables students to build a telescope that is similar to Galileo’s.
Build a telescope similar to a pinhole camera
50 mm lens for clear viewing
Easy assembly and simple instructions
Engage students in a hands-on exploration of astronomy and optics with this classroom-friendly kit. Designed for group learning, it allows students to construct a simple refracting telescope and experiment with its components—no optical bench required. The finished telescope delivers clear, sharp images, making it perfect for beginners eager to observe the night sky. Inspired by Galileo’s early designs, this kit demonstrates how even basic instruments can lead to remarkable discoveries.
Delivery information: The kit includes 10 inner and outer tubes, plastic eye pieces lenses, foam holders, cardboard spaces/washer for eyepiece lenses, red plasic caps, and 1 set of instructions and activities. The Single kit contains everything needed to build one telescope.
Spécifications
- Désignation:Refracting telescope, single