Motic BA210E LED Compound Microscopes
Product Details & Documents
The BA210Elite model introduces superior improvements both in optical and mechanical performance. Meant to be used in Educational life sciences, Medical and a variety of Biological applications, the BA210E is designed for ease of use and longevity due to Motic's complete understanding of the educational market.
- Widefield high eyepoint eyepiece N-WF10×/20 mm with diopter
- Adjustment on both eyepieces, with rubber eyecup (paired)
- Reversed quadruple revolving nosepiece
- Coaxial coarse and fine focusing adjustment
- Built in low position coaxial mechanical stage (right-hand control)
- Focusable Abbe condenser N.A. 1.25 with iris diaphragm and slot
The BA210E model introduces student-proof features both in optical and mechanical aspects. The new rackless stage concept without prominent gear rack allows even more convenient use of the x/y movement, while increasing user safety in teaching situations.
For full freedom of illumination options, the new model BA210E has an easy interchangeability of its 6 V/30 W Halogen bulb with LED modules of different colour temperatures (4500K, 6000K). Extended life-time of LEDs here is combined with cost saving aspects. Light consuming contrast methods like Phase contrast, Polarization or Dark field can easily performed.
Ordering information: Contact your Avantor representative to learn more about this and other motic solutions. All motic products come with a one-year electrical and five-year mechanical warranty.
Delivery information: The BA210E Phase package option (76163-614) includes upright compound microscope, trinocular head, widefield high eyepoint 10× eyepieces, CCIS EC Plan achromat/phase objectives (PL4×, PL100× S-Oil, Ph10X), rackless stage, NA 1.25 abbe condenser, phase slider, phase centering telescope, 3WLED module.
Specifications
- Désignation:BA210E Phase package