Coriolis® Air Sampler Accessories, Bertin Instruments
Product Details & Documents
Coriolis® air sampler consumables are part of the innovative dry cyclonic technology, which aspirates airborne particles and centrifuges them in a cone.
- Single use consumables
- Ensure optimal collection efficiency for Coriolis® air samplers
The separation of the airborne particles from the air flow is due to the air flow rate, the air intake geometry, the design of the cones, and the collection liquid (surfactant in low concentration). The collected particles or microorganisms can then be recovered by rinsing the consumables with an appropriate buffer solution, so the sample is compatible with multiple downstream analysis options (NGS, qPCR, culture).
Specifications
- Désignation:Sterile cone, liquid dose
- Matière:Polycarbonate