VWR® Macro Pipette Tips | Avantor
VWR® Macro Pipette Tips
Macro pipette tips, 5000 - 10000 µl, +, 143.8 mm, Bleu, Socorex, Finn, Rack of 25 Tips
Catalog # 76635-662
Supplier:   VWR International
VWR® Macro Pipette Tips
Catalog # 76635-662
Supplier:  VWR International
VWR® Macro Pipette Tips
VWR® Macro Pipette Tips
Macro pipette tips, 5000 - 10000 µl, +, 143.8 mm, Bleu, Socorex, Finn, Rack of 25 Tips
Price
$19.47
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pack of 25 Case of 250
Product Details & Documents
Improved accuracy over serological pipettes.
  • Tip length facilitates access into bottles, cell culture flasks, and tubes
Universal fit for compatibility with most major pipettor brands.
Certifications: Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free. Sterile product is certified pyrogen-free.
Documents
Spécifications
  • Volume:
    5000 - 10000 µl
  • Longueur:
    143.8 mm
  • Tip Color:
    Bleu
  • Packed:
    Rack of 25 Tips
  • Sterile:
    +
  • Tip Style:
    Macro pipette tips
  • For Use With:
    Socorex, Finn