VWR® Macro Pipette Tips
Macro pipette tips, 5000 - 10000 µl, +, 143.8 mm, Bleu, Socorex, Finn, Rack of 25 Tips
Supplier: VWR International
Jump to:
Product Details & Documents
Improved accuracy over serological pipettes.
- Tip length facilitates access into bottles, cell culture flasks, and tubes
Universal fit for compatibility with most major pipettor brands.
Certifications: Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free. Sterile product is certified pyrogen-free.
Documents
Certificates
Recherche par numéro de lotSpécifications
- Volume:5000 - 10000 µl
- Longueur:143.8 mm
- Tip Color:Bleu
- Packed:Rack of 25 Tips
- Sterile:+
- Tip Style:Macro pipette tips
- For Use With:Socorex, Finn
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.