VWR® Macro Pipette Tips
Macro pipette tips, 1000 - 5000 µl, +, 123.6 mm, Clear, Gilson, Rainin Classic and other universal-style pipettors, Rack of 50 Tips
Supplier: VWR International
Jump to:
Product Details & Documents
Improved accuracy over serological pipettes.
- Tip length facilitates access into bottles, cell culture flasks, and tubes
Universal fit for compatibility with most major pipettor brands.
Certifications: Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free. Sterile product is certified pyrogen-free.
Documents
Certificates
Recherche par numéro de lotSpécifications
- Volume:1000 - 5000 µl
- Longueur:123.6 mm
- Tip Color:Clear
- Packed:Rack of 50 Tips
- Sterile:+
- Tip Style:Macro pipette tips
- For Use With:Gilson, Rainin Classic and other universal-style pipettors