Trinocular Inverted Microscopes with Moticam, Camera Bundle, AE2000
Product Details & Documents
Designed for routine-lab or clinical work, as well as research requirements in pharmaceutical laboratories or universities, the AE2000 is more than capable of fulfilling the expectations of its users. Superb optical quality can be expected from a brand new optical series of both bright field and phase contrast lenses, following Motic ́s proven successful CCIS® Infinity Optical System.
- AE2000 Inverted
- Trinocular head Siedentopf type 45° inclined (light split: ²⁰/₈₀)
- Widefield high eyepoint eyepiece 10×/20 mm (paired), focusable
- Side facing quadruple nosepiece
- Moticam BMH4000X
- 1920×1080 HDMI monitor with attachable microscopy camera with 4K (3840×2160) resolution in SD card
- 4K through HDMI
- 20 fps at 3840×2160 through USB connection
- 12 V power adapter, CS ring, Motic 4-dot calibration slide, dust cap, manual, 12 mm focusable lens, 30 mm and 38 mm eyepiece adapters, macro tube, macro calibration card
- Motic Images Plus 3.1 for Windows/macOS
- Moticam BTI10
- 10" LCD android tablet with 1280×800 screen resolution
- Connection Wi-Fi (2.4 and 5 GHz), Ethernet
- WiFi resolution 4 MP (2592×1520 pixels) captured resolution
- WiFi output (802.1 a/b/g/n/ac supporting both hot-spot mode or using existing WiFi network)
- Preloaded optimized MotiConnect app
The Moticam BMH4000X is a camera that was designed for ease of use and the ability to output fast live images at 4K (3840×2160) pixels in full HD. Unlike other cameras, the Moticam BMH4000X can be connected to a computer or through HDMI. Simply plug the camera through the HDMI port and display images with the use of the onboard software and a USB mouse through the built-in 11.6" LCD screen, or connect your camera through USB to a Windows, Mac or Linux computer.
The Moticam BTI10 unlike other Moticams is combined with a 4.0 MP and a 10" android tablet that will deliver fast live images up to 30 fps on any trinocular microscope with the right C-mount adapter. With the convenience of a detachable tablet and WiFi image data transfer, the flexibility to capture images and bring your images anywhere has never been so easy.
Ordering information: Contact your Avantor representative to learn more about this and other motic solutions. All motic products come with a one-year electrical and five-year mechanical warranty.
Delivery information: Digital camera bundle comes with AE2000 Inverted Trinocular, Moticam BMH4000 or Moticam BTI10, compatible C-mount adapter.
Spécifications
- Désignation:AE2000 trinocular inverted microscope with Moticam BMH4000