Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Capacitors

Capacitors

Capacitor, 25 V, 220 µF
Catalog # 470347-904
Supplier #:  25V220UFPK10
Capacitors
Catalog # 470347-904
Capacitors

Capacitor, 25 V, 220 µF

Clearance
$0.90
Sale

Valid until 31/12/2026

Log in for availability
Each Pack of 10
Product Details & Documents

Compact electric storage for circuits.

  • 25 V
  • Radial design
  • Connects easily to bred boards

Capacitors are useful when you need to store a charge and release it in a circuit. These consist of two parallel plate capacitors that are arranged in a rolled-up design to minimize its volume.

Specifications
  • Désignation:
    Capacitor, 25 V, 220 µF