Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Capacitors | Avantor

Capacitors

Capacitor, 25 V, 100 µF
Catalog # 470347-902
Fournisseur #:  25V100UFPK10
Capacitors
Catalog # 470347-902
Capacitors

Capacitor, 25 V, 100 µF

Liquidation
$0.85
Vente

Valable jusqu'au 31/12/2026

Log in pour la disponibilité
Each Pack of 10
Product Details & Documents

Compact electric storage for circuits.

  • 25 V
  • Radial design
  • Connects easily to bred boards

Capacitors are useful when you need to store a charge and release it in a circuit. These consist of two parallel plate capacitors that are arranged in a rolled-up design to minimize its volume.

Spécifications
  • Désignation:
    Capacitor, 25 V, 100 µF
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.