CanDo® Exercise Bands | Avantor
CanDo® Exercise Bands
XXX-Heavy exercise bands, Gold, 5", 25 yd.
Catalog # 470348-064
Fournisseur #:  10-5637
CanDo® Exercise Bands
Catalog # 470348-064
CanDo® Exercise Bands
CanDo® Exercise Bands
XXX-Heavy exercise bands, Gold, 5", 25 yd.
Price
141,60 $
Log in pour la disponibilité
Each
Product Details & Documents
Bulk rolls for easy length adjustment.
  • Latex-free
  • Lightweight, compact, and portable
  • Durable
CanDo® resistive band exerciser rolls are packaged in a dispenser box. Simply unroll desired length from dispenser box and cut with scissors. CanDo® high-quality synthetic latex-free exercise bands have been used for physical therapy and athletic training by clinics, fitness centers, and individuals since 1987. It has become a well known staple of many physical therapy programs. The latex-free formula has been specially created to mimic the stretch characteristics of latex bands. CanDo® exercise bands are an economic solution for upper and lower body exercise. Effective when used alone, or with handles and anchors. Color-coded according to the exercise band's resistance. CanDo® is licensed to use the Thera-Band color sequence so resistance level is easily identified. Available in 8 levels of resistance for progressive exercise.
Spécifications
  • Color:
    Gold
  • Désignation:
    XXX-Heavy exercise bands
  • Longueur:
    25 yd.
  • Width:
    5"