CanDo® Exercise Bands
Product Details & Documents
Bulk rolls for easy length adjustment.
- Latex-free
- Lightweight, compact, and portable
- Durable
CanDo® resistive band exerciser rolls are packaged in a dispenser box. Simply unroll desired length from dispenser box and cut with scissors. CanDo® high-quality synthetic latex-free exercise bands have been used for physical therapy and athletic training by clinics, fitness centers, and individuals since 1987. It has become a well known staple of many physical therapy programs. The latex-free formula has been specially created to mimic the stretch characteristics of latex bands. CanDo® exercise bands are an economic solution for upper and lower body exercise. Effective when used alone, or with handles and anchors. Color-coded according to the exercise band's resistance. CanDo® is licensed to use the Thera-Band color sequence so resistance level is easily identified. Available in 8 levels of resistance for progressive exercise.
Specifications
- Color:Black
- Désignation:X-Heavy exercise bands
- Longueur:25 yd.
- Width:5"