Estes Wizard Model Rockets
Product Details & Documents
Intermediate-level rocket with laser-cut balsa fins and streamer recovery system.
- Intermediate-level build
- Requires standard size 18 mm A, B, or C engines (A or B recommended)
- Max. altitude 488 m (1600')
Looking for a magical experience? Are your model rocketry skills ready to level-up? The Estes wizard rocket is exactly what you need! This high-performance rocket is easy to build and known for casting spells on all who own it.
With powerful Estes model rocket engines, this light-weight rocket can reach heights of up to 1600 feet in the blink of an eye. Its laser-cut balsa wood fins still make assembly easy, and its streamer recovery system helps it descend efficiently and safely from high altitudes. This sleek rocket is fun to build and thrilling to fly.
: Launch system and rocket engines available separately.
Single kit includes all parts needed to assemble (1) model rocket. Some tools and glue may be required.
Bulk pack includes: (12) single kits.
Spécifications
- Désignation:Wizard model rocket kit (bulk pack)
- Packaging:Pack of 12
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



