Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
VWR® ExoCleanUp FAST

VWR® ExoCleanUp FAST

ExoCleanUp FAST, 1× 0.2 ml, 100
  76620-568
VWR® ExoCleanUp FAST
  76620-568
 :  
VWR® ExoCleanUp FAST
VWR® ExoCleanUp FAST

ExoCleanUp FAST, 1× 0.2 ml, 100

 :  
 
$198.90
   
Each
 

ExoCleanUp FAST PCR reagent is a one-step PCR clean-up reagent for optimal sequencing results, consisting of a balanced combination of a heat-labile exonuclease I (HL-ExoI) and a recombinant shrimp alkaline phosphatase (rSAP).

  • Designed to clean-up PCR products in 5 minutes
  • No need for spin columns or magnetic beads
  • Treatment improves downstream applications, such as DNA sequencing and SNP analysis

Treatment of amplified PCR products with this reagent helps to remove residual primers and single-stranded DNA, and inactivates excess dNTPs by dephosphorylation. After enzymatic treatment at 37 °C for a minimum of 2 minutes, this reagent is completely inactivated by heating at 80 °C for a minimum of 3 minutes.

Caution: For Research Use Only. Not for use in diagnostic procedures.

 

 

  • Désignation:
    ExoCleanUp FAST
  • No. of Reactions:
    100
  • Volume:
    1× 0.2 ml

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.