Large Pathology Containers, Mopec | Avantor
Large Pathology Containers, Mopec
1 gal., Large pathology containers
Catalog # 76579-420
Supplier:   Mopec
Fournisseur #:  BG615
Large Pathology Containers, Mopec
Catalog # 76579-420
Supplier:  Mopec
Large Pathology Containers, Mopec
Large Pathology Containers, Mopec
1 gal., Large pathology containers
Price
$63.02
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Mopec’s screw top pathology containers are made from a high-quality, clarified polypropylene material. The screw-top lid creates a moisture and tamper-resistant closure that increases safety for laboratory employees and helps protect pathology specimens during transportation. Features an attached handle for easy transportation. Available in multiple sizes.
  • Made from a high-quality, clarified polypropylene material
  • Screw-top lid creates a moisture and tamper-resistant closure
  • Features an attached handle for easy transportation
Sold individually.
Spécifications
  • Capacité:
    1 gal.
  • Désignation:
    Large pathology containers