Large Pathology Containers, Mopec
2 gal., Large pathology containers
  76579-430
 :   Mopec
 :  BG625
Large Pathology Containers, Mopec
  76579-430
 :  Mopec
Large Pathology Containers, Mopec
Large Pathology Containers, Mopec
2 gal., Large pathology containers
$112.09
Each
Mopec’s screw top pathology containers are made from a high-quality, clarified polypropylene material. The screw-top lid creates a moisture and tamper-resistant closure that increases safety for laboratory employees and helps protect pathology specimens during transportation. Features an attached handle for easy transportation. Available in multiple sizes.
  • Made from a high-quality, clarified polypropylene material
  • Screw-top lid creates a moisture and tamper-resistant closure
  • Features an attached handle for easy transportation
Sold individually.
  • Capacité:
    2 gal.
  • Désignation:
    Large pathology containers