Quantum Levitation Extended Duration Levitators | Avantor
Quantum Levitation Extended Duration Levitators
Quantum extended duration levitator, med
Catalog # 470346-490
Fournisseur #:  15-2
Quantum Levitation Extended Duration Levitators
Catalog # 470346-490
Quantum Levitation Extended Duration Levitators
Quantum Levitation Extended Duration Levitators
Quantum extended duration levitator, med
Price
$1,646.29
Log in pour la disponibilité
Each Pack of 1
Product Details & Documents
The quantum enhanced-duration levitators are the perfect tool for science demonstrations and visual display. Engineered with years of experience and knowhow in cryogenics, these levitators offer the optimal balance between levitation strength and levitation time.
  • Quantum extended duration levitator
  • Plastic tweezers
  • User manual
These new levitators offer 2 to 3× times longer levitation times compared to our standard ones. This means you’ll have more time to explain and discuss the physics or just appreciate their magic instead of constantly refilling with liquid nitrogen.
Spécifications
  • Désignation:
    Quantum extended duration levitator, med