Eisco Plastic Scoops
Plastic scoops, 500 ml
Catalog # 470345-896
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
500ML PLASTIC SCOOP PK6 Catalog #470345-897
Product 470345-897 is a replacement for 470345-896 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Pack of 12 or 6 weighing scoops with labeled handles and flat bottoms.
- Made of heat- and chemical-resistant polypropylene plastic
- Scoop handles are labeled with capacity for easy identification
- Flat bottom design allows scoops to rest upright for weighing and filling
Autoclavable and dishwasher safe for easy cleaning.
Specifications
- Capacité:500 ml
- Désignation:Plastic scoops