Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Eisco Plastic Filter Funnels

10, 100 mm, 80 mm
Catalog # 470345-846
Supplier:   Wards
 :  CH0454D10
Eisco Plastic Filter Funnels
Catalog # 470345-846
Supplier:  Wards
Eisco Plastic Filter Funnels
Eisco Plastic Filter Funnels

10, 100 mm, 80 mm

Price
$20.65
Log in pour la disponibilité
Each
Product Details & Documents

Chemically-resistant filter funnels made of high quality polypropylene plastic.

  • Excellent resistance against acids, alcohols, aldehydes, bases, and hydrocarbons
  • Useful for separating solids from liquids, as well as for easier transfers of liquids to narrow mouthed containers
  • Autoclavable and microwavable

 : Pack of 1 or 10.

Spécifications
  • Ø:
    100 mm
  • Taille:
    10
  • Stem Length:
    80 mm