Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Eisco® Hooked Weights, Iron | Avantor

Eisco® Hooked Weights, Iron

500 g
Catalog # 470345-752
Supplier:   Wards
Fournisseur #:  HKW500
Eisco® Hooked Weights, Iron
Catalog # 470345-752
Supplier:  Wards
Eisco® Hooked Weights, Iron
Eisco® Hooked Weights, Iron

500 g

Price
$20.30
Log in pour la disponibilité
Each
Product Details & Documents

Hooked Iron Weights with cylindrical bodies stamped with weight, cone-shaped tops, and bottom bar and slot to hang additional weights.

  • Cylindrical with cone-shaped top
  • Integral hook for easy lifting and hanging
  • Slotted bottom with a bar for hanging additional hooked weights underneath (excluding 10 g weight)
  • Each piece is stamped with weight for easy identification
  • Ideal as additional or spare part for hooked weight sets

Single hooked weights made of high-quality iron. Cylindrical with cone-shaped top. Features hook for easy lifting and hanging. Bottom slot allows for other hooked weights to be attached in tandem. Stamped with weight for easy identification.

Spécifications
  • Weight:
    500 g