Eisco® Hooked Weights, Brass
200 g
Catalog # 470345-450
Supplier: Wards
Fournisseur #: PHHWBR5
Jump to:
Product Details & Documents
Hooked stainless brass weights with cylindrical bodies and stamped with weight for easy identification.
- Made from high-quality brass
- Integral hook for easy lifting and hanging
- Each piece stamped with weight for easy identification
- Ideal as additional or spare part for hooked weight sets
Cylindrical single weights made of high-quality brass feature a hook for easy lifting and hanging. Stamped with weight for easy identification. Great as replacements for missing pieces or as spares.
Caution: Warning: cancer and reproductive harm (www.p65warnings.ca.gov).
Spécifications
- Weight:200 g