Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Eisco Plastic Conical Flasks with Screw Cap | Avantor

Eisco Plastic Conical Flasks with Screw Cap

250 ml, Translucent
Catalog # 470345-050
Supplier:   Wards
Fournisseur #:  CH0431B
Eisco Plastic Conical Flasks with Screw Cap
Catalog # 470345-050
Supplier:  Wards
Eisco Plastic Conical Flasks with Screw Cap
Eisco Plastic Conical Flasks with Screw Cap

250 ml, Translucent

Price
$8.90
Log in pour la disponibilité
Each
Product Details & Documents

Polypropylene conical flasks with screw caps and no graduations.

  • Made from high-quality polypropylene
  • Highly chemical resistant, microwavable and autoclavable at 121 °C
  • Tight-fitting screw caps for transportation and storage of contents
  • Wide neck for easy filling and cleaning
  • Translucent body makes it easy to see liquid level in the flask

Conical flasks with a wide neck and a screw cap. The natural translucent color makes it easy to see any liquid level in the bottle. Made from easy-to-clean high-quality polypropylene. Highly chemical resistant, microwavable and autoclavable at 121 °C.

Spécifications
  • Capacité:
    250 ml
  • Color:
    Translucent