Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Eisco Plastic Filter Funnels | Avantor

Eisco Plastic Filter Funnels

10, 50 mm, 50 mm
Catalog # 470344-828
Eisco Plastic Filter Funnels
Catalog # 470344-828
Supplier:  
Eisco Plastic Filter Funnels
Eisco Plastic Filter Funnels

10, 50 mm, 50 mm

Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
FUNNEL 9MM STEM DIAM PP PK24 Catalog #83008-720

Le produit 83008-720 est similaire à 470344-828 mais le fabricant et les spécifications peuvent varier.

Product Details & Documents

Chemically-resistant filter funnels made of high quality polypropylene plastic.

  • Excellent resistance against acids, alcohols, aldehydes, bases, and hydrocarbons
  • Useful for separating solids from liquids, as well as for easier transfers of liquids to narrow mouthed containers
  • Autoclavable and microwavable

Packaging: Pack of 1 or 10.

Spécifications
  • Ø:
    50 mm
  • Taille:
    10
  • Stem Length:
    50 mm