Eisco Plastic Scoops
Plastic scoops, 2 ml
Catalog # 470344-582
Eisco Plastic Scoops
Catalog # 470344-582
Supplier:  
Eisco Plastic Scoops
Eisco Plastic Scoops
Plastic scoops, 2 ml
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
2ML PLASTIC SCOOP PK12 Catalog #470344-583
Product 470344-583 is a replacement for 470344-582 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Pack of 12 or 6 weighing scoops with labeled handles and flat bottoms.
  • Made of heat- and chemical-resistant polypropylene plastic
  • Scoop handles are labeled with capacity for easy identification
  • Flat bottom design allows scoops to rest upright for weighing and filling
Autoclavable and dishwasher safe for easy cleaning.
Spécifications
  • Capacité:
    2 ml
  • Désignation:
    Plastic scoops