Eisco Plastic Scoops
Plastic scoops, 2 ml
Catalog # 470344-582
Jump to:
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
2ML PLASTIC SCOOP PK12 Catalog #470344-583
Product 470344-583 is a replacement for 470344-582 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Pack of 12 or 6 weighing scoops with labeled handles and flat bottoms.
- Made of heat- and chemical-resistant polypropylene plastic
- Scoop handles are labeled with capacity for easy identification
- Flat bottom design allows scoops to rest upright for weighing and filling
Autoclavable and dishwasher safe for easy cleaning.
Spécifications
- Capacité:2 ml
- Désignation:Plastic scoops