Eisco Fixed Volume Micropipettes
Product Details & Documents
Factory-calibrated, color-coded Fixed Volume Micropipettes with tip ejector, PVDF cone, and body grip conform to DIN 12650 and EN ISO 8655 standards.
- Fixed volume micropipettes
- Features a soft and lightweight operation with an ergonomic, soft touch tip ejector designed to minimize hand fatigue, reduce thumb stress and to increase comfort with prolonged use
- Fully autoclavable with a durable, autoclavable PVDF tip cone and a slip-resistant thermoplastic elastomer (TPE) body grip to minimize the transfer of body heat to the micropipette resulting in high accuracy even with continuous use
- Entire series of micropipettes features color coded plungers for easy identification
- Calibration conforms to DIN 12650 and EN ISO 8655 standard ensuring high accuracy and precision
- Calibration report enclosed with every micropipette
High performance fixed volume micropipettes have a simple design which allows for easy use. Features soft and lightweight operation with an ergonomic, soft touch tip ejector designed to minimize hand fatigue, reduce thumb stress and to increase comfort with prolonged use.
Fully autoclavable with a durable, autoclavable PVDF tip cone and a slip-resistant thermoplastic elastomer (TPE) body grip to minimize the transfer of body heat to the micropipette resulting in high accuracy even with continuous use.
The entire series of Eisco Fixed Volume Micropipettes features color coded plungers for easy identification. Calibration conforms to DIN 12650 and EN ISO 8655 standard ensuring high accuracy and precision. Calibration report enclosed with every micropipette.
`
Spécifications
- Volume:20 µl
- Désignation:Micropipette
- Color Code:Yellow