Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Eisco® Hooked Weights, Iron

Eisco® Hooked Weights, Iron

200 g
Catalog # 470344-386
Supplier:   Wards
Supplier #:  HKW200
Eisco® Hooked Weights, Iron
Catalog # 470344-386
Supplier:  Wards
Eisco® Hooked Weights, Iron
Eisco® Hooked Weights, Iron

200 g

Price
$17.25
Log in pour la disponibilité
Each
Product Details & Documents

Hooked Iron Weights with cylindrical bodies stamped with weight, cone-shaped tops, and bottom bar and slot to hang additional weights.

  • Cylindrical with cone-shaped top
  • Integral hook for easy lifting and hanging
  • Slotted bottom with a bar for hanging additional hooked weights underneath (excluding 10 g weight)
  • Each piece is stamped with weight for easy identification
  • Ideal as additional or spare part for hooked weight sets

Single hooked weights made of high-quality iron. Cylindrical with cone-shaped top. Features hook for easy lifting and hanging. Bottom slot allows for other hooked weights to be attached in tandem. Stamped with weight for easy identification.

Spécifications
  • Weight:
    200 g
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.