Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Eisco Plastic Conical Flasks with Screw Cap | Avantor

Eisco Plastic Conical Flasks with Screw Cap

500 ml, Translucent
Catalog # 470344-122
Supplier:   Wards
Fournisseur #:  CH0431C
Eisco Plastic Conical Flasks with Screw Cap
Catalog # 470344-122
Supplier:  Wards
Eisco Plastic Conical Flasks with Screw Cap
Eisco Plastic Conical Flasks with Screw Cap

500 ml, Translucent

Price
$16.05
Log in pour la disponibilité
Each
Product Details & Documents

Polypropylene conical flasks with screw caps and no graduations.

  • Made from high-quality polypropylene
  • Highly chemical resistant, microwavable and autoclavable at 121 °C
  • Tight-fitting screw caps for transportation and storage of contents
  • Wide neck for easy filling and cleaning
  • Translucent body makes it easy to see liquid level in the flask

Conical flasks with a wide neck and a screw cap. The natural translucent color makes it easy to see any liquid level in the bottle. Made from easy-to-clean high-quality polypropylene. Highly chemical resistant, microwavable and autoclavable at 121 °C.

Spécifications
  • Capacité:
    500 ml
  • Color:
    Translucent
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.