Eisco High-Performance Variable Volume Micropipettes
5 - 50 µl, 0.50 µl, Yellow, 1
470344-014
: Wards
: VVM3
Variable volume micropipettes are fully autoclavable and feature a simple ergonomic design for comfort, color coded plungers, and calibration conforming to DIN ISO standards.
- Soft and lightweight operation with an ergonomic, soft touch tip ejector designed to minimize hand fatigue, reduce thumb stress and to increase comfort with prolonged use
- Fully autoclavable with a durable, autoclavable PVDF tip cone
- Slip-resistant thermoplastic elastomer (TPE) body grip to minimize body heat transfer to the micropipette during long use, resulting in higher accuracy
- Color coded plungers for easy identification
- Calibration conforms to DIN 12650 & EN-ISO 8655 standard ensuring high accuracy and precision - Calibration report enclosed with every micropipette
High performance variable volume micropipettes have a simple design which allows for easy adjustment and reading of increments.
- Volume:5 - 50 µl
- Increments:0.50 µl
- No. of Channels:1
- Color Code:Yellow