Eisco Plastic Scoops
Plastic scoops, 5 ml
470343-956
Pack of 12 or 6 weighing scoops with labeled handles and flat bottoms.
- Made of heat- and chemical-resistant polypropylene plastic
- Scoop handles are labeled with capacity for easy identification
- Flat bottom design allows scoops to rest upright for weighing and filling
Autoclavable and dishwasher safe for easy cleaning.
- Capacité:5 ml
- Désignation:Plastic scoops