Eisco® Hooked Weights, Iron
10 g
470343-914
: Wards
: HKW10G
Hooked Iron Weights with cylindrical bodies stamped with weight, cone-shaped tops, and bottom bar and slot to hang additional weights.
- Cylindrical with cone-shaped top
- Integral hook for easy lifting and hanging
- Slotted bottom with a bar for hanging additional hooked weights underneath (excluding 10 g weight)
- Each piece is stamped with weight for easy identification
- Ideal as additional or spare part for hooked weight sets
Single hooked weights made of high-quality iron. Cylindrical with cone-shaped top. Features hook for easy lifting and hanging. Bottom slot allows for other hooked weights to be attached in tandem. Stamped with weight for easy identification.
- Weight:10 g