Density Cubes with Hook
Product Details & Documents
Cubes have 2 cm(0.78") sides with a hook for density investigation and specific gravity activities. Element stamp indicates material.
- Single, solid density cube with hook and stamped with element for easy identification
- Measures 2 cm(0.78") cubed, including the hook, the cube measures 1.5" in height
- Excellent for physics classrooms
- Density Cubes have a wide range of educational applications for all grade levels, including density investigation, specific heat and specific gravity exploration
Since these cubes are equipped with hooks, they can be easily lifted from water. Additionally, the hooks allow the cube to be easily massed using a spring scale. Cubes have a mass between 50 g and 400 g, so a 5.0 to 1.0 Newton spring scale will work perfectly.
The Eisco labs density investigation series is designed to help students identify and figure out mass, understand the relationship between mass/volume/density, understand specific heat and specific gravity, and other introductory physics and chemistry concepts.
Caution: Due to the nature of raw metals, some oxidation or corrosion due to humidity and storage may occur. Simply scuff any oxidation off to make cube look brand new again.
Designed to be used in an educational setting and are not fabricated using ultra high precision machining. Expect tolerances in the ±5% range of any stated dimensions (although they are generally much tighter).
Specifications
- Désignation:Denisty cube, lead (Pb)