Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Eisco Eudiometer Tubes | Avantor

Eisco Eudiometer Tubes

50 ml, Eudiometer tubes, 0.1 ml graduations, Borosilicate glass 3.3
Catalog # 470343-752
Supplier:   Wards
Fournisseur #:  CH0499
Eisco Eudiometer Tubes
Catalog # 470343-752
Supplier:  Wards
Eisco Eudiometer Tubes

50 ml, Eudiometer tubes, 0.1 ml graduations, Borosilicate glass 3.3

Price
$66.25
Log in pour la disponibilité
Each
Product Details & Documents

Eudiometer tubes with platinum elecrodes for measuring gas volume changes during chemical reactions.

  • Made of mechanically strong and chemical resistant Borosilicate 3.3 glass
  • Contains two platinum electrodes on the sealed end of tube
  • Easy-to-read white graduations

Eudiometer tubes made of high quality Borosilicate glass. Useful for measuring volume of gas produced or consumed in a chemical reaction.

Spécifications
  • Capacité:
    50 ml
  • Désignation:
    Eudiometer tubes, 0.1 ml graduations
  • Matière:
    Borosilicate glass 3.3