Eisco® Hooked Weights, Brass | Avantor
Eisco® Hooked Weights, Brass
500 g
Catalog # 470343-724
Supplier:   Wards
Fournisseur #:  PHHWBR6
Eisco® Hooked Weights, Brass
Catalog # 470343-724
Supplier:  Wards
Eisco® Hooked Weights, Brass
Eisco® Hooked Weights, Brass
500 g
Price
$82.95
Log in pour la disponibilité
Each
Product Details & Documents
Hooked stainless brass weights with cylindrical bodies and stamped with weight for easy identification.
  • Made from high-quality brass
  • Integral hook for easy lifting and hanging
  • Each piece stamped with weight for easy identification
  • Ideal as additional or spare part for hooked weight sets
Cylindrical single weights made of high-quality brass feature a hook for easy lifting and hanging. Stamped with weight for easy identification. Great as replacements for missing pieces or as spares.
Caution: Warning: cancer and reproductive harm (www.p65warnings.ca.gov).
Spécifications
  • Weight:
    500 g