Klercide™ mop wipe frame, stainless steel
Mop wipe frame, incl. one head and one handle, Stainless steel
Catalog # 76554-276
Supplier: ECOLAB LIFE SCIENCES
Fournisseur #: 10056359
Jump to:
Product Details & Documents
This stainless steel mop frame features a stiff movable flat top.
- Fully autoclavable
- Made from stainless steel with stiff, moveable flat mop head frame
- Designed for use with Klercide™ pre-saturated mop wipes
- Supplied Gamma irradiated
Option of adding Klercide™ silicone pad for better contact on uneven surfaces.
Spécifications
- Désignation:Mop wipe frame, incl. one head and one handle
- Matière:Stainless steel