Klercide™ mop wipe frame, stainless steel | Avantor
Klercide™ mop wipe frame, stainless steel
Mop wipe frame, incl. one head and one handle, Stainless steel
Catalog # 76554-276
Fournisseur #:  10056359
Klercide™ mop wipe frame, stainless steel
Catalog # 76554-276
Klercide™ mop wipe frame, stainless steel
Klercide™ mop wipe frame, stainless steel
Mop wipe frame, incl. one head and one handle, Stainless steel
Price
$1,011.57
Log in pour la disponibilité
Each
Product Details & Documents
This stainless steel mop frame features a stiff movable flat top.
  • Fully autoclavable
  • Made from stainless steel with stiff, moveable flat mop head frame
  • Designed for use with Klercide™ pre-saturated mop wipes
  • Supplied Gamma irradiated
Option of adding Klercide™ silicone pad for better contact on uneven surfaces.
Spécifications
  • Désignation:
    Mop wipe frame, incl. one head and one handle
  • Matière:
    Stainless steel